Order Entry
Austria
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

Bottles, 2
Catalog # 715-3224
Corrosive
Supplier:   WTW
Sodium hydroxide
Catalog # 715-3224
Supplier:  WTW
CAS Number:  1310-73-2
Sodium hydroxide

Bottles, 2

CAS Number:  1310-73-2
Price
70,00 €
Log in for availability
1 * 2 ST
Product Details & Documents

Sodium hydroxide pellets is used as the carbon dioxide absorbent for BOD measurements

  • Product category: PC21 laboratory chemicals
  • Process category: PROC15 use as laboratory reagent
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:319 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pack type:
    Bottles
  • Pk:
    2

Specifications