Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Corrosive
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Total N (Nitrogen):
    Max. 5 ppm
  • CO3 (as Na2CO3):
    Max. 1.0 %
  • Cl (Chloride):
    Max. 50 ppm
  • PO4 (Phosphate):
    Max. 20 ppm
  • SiO4 (as SiO2):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Al (Aluminium):
    Max. 10 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 0.1 %
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

Frequently Bought Together