Sodium hydroxide 95.0-100.5%, pellets, GenAR® NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets F.C.C. Requirements:—
- GMP Manufactured Product:—
- Meets E.P. Chemical Specifications:—
- Meets N.F. Requirements:—
- Food GMP Manufactured Product:—
- Meets J.P. Chemical Specifications:—
- Meets B.P. Chemical Specifications:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- Bulk Food Chemical:95.0 - 100.5 %
- NF - Total Alkali:95.0 - 100.5 %
- NF - Content of Sodium:54.0 - 59.8 %
- NF - Insoluble Substances and Organic Matter:Passes Test
- NF - Identification A (Sodium):Passes Test
- NF - Identification B (pH):≥11.0
- NF - Potassium (K):≤0.5 %
- NF/FCC - Carbonate (as Na₂CO₃):≤3.0 %
- FCC - Assay (Total alkali as NaOH):95.0 - 100.5 %
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Mercury (Hg)(mg/kg):≤0.1
- FCC - Arsenic (As)(mg/kg):≤3
- FCC - Identification:Passes Test
- FCC - Insoluble Substances and Organic Matter:Passes Test
- BP/EP - Assay (Total alkali as NaOH):97.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Carbonate (as Na₂CO₃):≤2.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- JP - Assay:95.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.050 %
- JP - Heavy Metals (as Pb):≤30 ppm
- JP - Potassium (K):Passes Test
- JP - Carbonate (as Na₂CO₃):≤2.0 %
- JP - Mercury (Hg):Passes Test
- Endotoxin Concentration (EU/g):≤5
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_MACR7772-06 spec_MACR7772-25 spec_MACR7772-28 spec_MACR7772-30 spec_MACR7772-88
Product specification
Frequently Bought Together