Order Entry
Austria
ContactUsLinkComponent
 
Sodium hydroxide 95.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
 
undefined
Sodium hydroxide 95.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
 

 

  • Environmentally Preferable:

 

 

  • GMP Manufactured Product:
  • Food GMP Manufactured Product:
  • Meets F.C.C. Requirements:
  • Meets N.F. Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • Bulk Food Chemical:
    95.0 - 100.5 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • NF - Appearance (white hygroscopic pellets):
    Passes Test
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥11.0
  • NF - Potassium (K):
    ≤0.5 %
  • NF - Sodium Carbonate (Na₂CO₃):
    ≤3.0 %
  • FCC - Assay (Total alkali as NaOH) (by acidimetry):
    95.0 - 100.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Insoluble Substances and Organic Matter:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Mercury (Hg):
    ≤0.1 mg/kg
  • FCC - Arsenic (As):
    ≤3 mg/kg
  • FCC - Sodium Carbonate (Na₂CO₃):
    ≤3.0 %

 

Frequently Bought Together