Order Entry
Austria
ContactUsLinkComponent
Sodium peroxide ≥95%, small beads ACS for Wurzschmitt-decomposition, Fluka™
Sodium peroxide ≥95%, small beads ACS for Wurzschmitt-decomposition, Fluka™
Catalog # 71883-1KG
CAS Number:  
Sodium peroxide ≥95%, small beads ACS for Wurzschmitt-decomposition, Fluka™
Catalog # 71883-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    95.0 - 105.0 %
  • Silver (Ag):
    Max. 5 ppm
  • Aluminium (Al):
    Max. 20 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 500 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 200 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 10 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 10 ppm
  • Thallium (Tl):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 20 ppm
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 10 ppm
  • Total N:
    Max. 30 ppm

Specification Test Results

About this item

Synonyms: di-Sodium peroxide
 :
 :77.98 g/mol
Merck Index:13,08728
Boiling Pt:Decomposes on heating
Boiling Pt: Dec. (on heating)
Melting Pt:660 °C
Density:2.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-209-4
UN:1504
ADR:5.1,I
 :GHS05
 :GHS03
CAS Number:1313-60-6
Formula:Na₂O₂
MDL Number:MFCD00003497