Orders LinkContactUsLinkComponent
Dishes, crystallising
Dishes, crystallising
Supplier:   KARTELL
Dishes, crystallising
Supplier:  KARTELL
Certificates
Dishes, crystallising
Dishes, crystallising
3 Options Available Below
Product Details & Documents
Crystallising dishes, made of PTFE.
  • Chemical, mechanical and heat resistance
  • Used safely for continuous service from −270 up to +250 °C
Suitable for dissolutions, digestions with acids and corrosive agents and suitable for food stuffs.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 100 ml
    54,48 mm
    64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,80 €
    65,80 €
    1 * 1 ST
       65,80 €
  • 25 ml
    35,64 mm
    38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,60 €
    46,60 €
    1 * 1 ST
       46,60 €
  • 350 ml
    62,22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €