Replacement parts for screw joints, BOLA
Supplier: Bohlender
Certificates
About this item
Made of glass-fibre reinforced PTFE.
- Temperature resistance: from –200 to +250 °C
- Very good chemical resistance
For fittings, valves and stopcocks.
Frequently Bought Together