Orders LinkContactUsLinkComponent
Counting chambers, Sedgewick
Counting chambers, Sedgewick
Supplier:   MARIENFELD
Counting chambers, Sedgewick
Supplier:  MARIENFELD
Certificates
Counting chambers, Sedgewick
Counting chambers, Sedgewick
2 Options Available Below
Product Details & Documents
Counting chambers according to Sedgewick for particles and microorganisms in transparent liquids.
A cell is ruled in a grid subdividing 1,0 ml into 1000 μl.
Delivery information: Supplied with a cover glass.
Product Family Options
Product Information
  • Depth of chamber
    W×D
    Availability
    Price
  • MARI0611300
    Cell counter (glass with electro-deposited chromium) with ground and chamfered edges
    Cell counter (glass with electro-deposited chromium) with ground and chamfered edges
    1 mm
    50×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 638,73 €
    638,73 €
    1 * 1 ST
       638,73 €
  • MARI0611400
    Cell counter (plastic)
    Cell counter (plastic)
    1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 133,33 €
    133,33 €
    1 * 1 ST
       133,33 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cover glass
    Cover glass
    MARI0336000
    Log in for availability
    1 * 1 ST - 40,33 €