Order Entry
Austria
Orders LinkContactUsLinkComponent
Water

Water

Marketsource
Supplier:   Roth Carl
Water
Supplier:  Roth Carl
Marketsource
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 13,40 €
    13,40 €
    1 * 1 L
       13,40 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 28,00 €
    28,00 €
    1 * 2,5 L
       28,00 €

    Specifications:
    Marketsource
    Marketsource
  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 43,50 €
    43,50 €
    1 * 5 L
       43,50 €

    Specifications:
    Marketsource
    Marketsource
  • 30 L
    Log in for availability
    Log in for availability
    1 * 30 L
    1 * 30 L - 176,00 €
    176,00 €
    1 * 30 L
       176,00 €

    Specifications:
    Marketsource
    Marketsource