Water, NORMATOM® for trace metal analysis
:7732-18-5
:
:H₂O
:MFCD00011332
:18.02 g/mol
:13,10098
:100 °C (1013 hPa)
:0 °C
:1 g/cm³ (4 °C)
:Ambient
:231-791-2
- Colouration:10 CO max.
- Ag (Silver):Max. 50 ppt
- Al (Aluminium):Max. 200 ppt
- As (Arsenic):Max. 50 ppt
- Au (Gold):Max. 50 ppt
- B (Boron):Max. 500 ppt
- Ba (Barium):Max. 50 ppt
- Be (Beryllium):Max. 50 ppt
- Bi (Bismuth):Max. 50 ppt
- Ca (Calcium):Max. 500 ppt
- Cd (Cadmium):Max. 50 ppt
- Ce (Cerium):Max. 20 ppt
- Co (Cobalt):Max. 50 ppt
- Cr (Chromium):Max. 50 ppt
- Cs (Cesium):Max. 50 ppt
- Cu (Copper):Max. 100 ppt
- Dy (Dysprosium):Min. 20 ppt
- Er (Erbium):Max. 20 ppt
- Eu (Europium):Max. 20 ppt
- Fe (Iron):Max. 300 ppt
- Ga (Gallium):Max. 50 ppt
- Gd (Gadolinium):Max. 20 ppt
- Ge (Germanium):Max. 50 ppt
- Hf (Hafnium):Max. 10 ppt
- Hg (Mercury):Max. 200 ppt
- Ho (Holmium):Max. 20 ppt
- In (Indium):Max. 50 ppt
- K (Potassium):Max. 300 ppt
- La (Lanthanum):Max. 20 ppt
- Li (Lithium):Max. 50 ppt
- Lu (Lutetium):Max. 20 ppt
- Mg (Magnesium):Max. 100 ppt
- Mn (Manganese):Max. 50 ppt
- Mo (Molybdenum):Max. 50 ppt
- Na (Sodium):Max. 500 ppt
- Nb (Niobium):Max. 20 ppt
- Nd (Neodymium):Max. 20 ppt
- Ni (Nickel):Max. 50 ppt
- Pb (Lead):Max. 50 ppt
- Pd (Palladium):Max. 50 ppt
- Pr (Praseodymium):Max. 20 ppt
- Pt (Platinum):Max. 50 ppt
- Rb (Rubidium):Max. 20 ppt
- Re (Rhenium):Max. 50 ppt
- Rh (Rhodium):Max. 50 ppt
- Ru (Ruthenium):Max. 50 ppt
- Sb (Antimony):Max. 50 ppt
- Sc (Scandium):Max. 20 ppt
- Se (Selenium):Max. 200 ppt
- Sm (Samarium):Max. 20 ppt
- Sn (Tin):Max. 50 ppt
- Sr (Strontium):Max. 50 ppt
- Ta (Tantalum):Max. 50 ppt
- Tb (Terbium):Max. 20 ppt
- Te (Tellurio):Max. 50 ppt
- Th (Thorium):Max. 10 ppt
- Ti (Titanium):Max. 50 ppt
- Tl (Thallium):Max. 50 ppt
- Tm (Thulium):Max. 20 ppt
- U (Uranium):Max. 10 ppt
- V (Vanadium):Max. 50 ppt
- W (Tungsten):Max. 100 ppt
- Y (Yttrium):Max. 10 ppt
- Yb (Ytterbium):Max. 20 ppt
- Zn (Zinc):Max. 500 ppt
- Zr (Zirconium):Max. 50 ppt
Frequently Bought Together