Order Entry
Austria
ContactUsLinkComponent
Multiple distributors for bottles, BOLA
Multiple distributors for bottles, BOLA
Catalog # BOHLD651-08
Supplier:  Bohlender
undefined
Multiple distributors for bottles, BOLA
Catalog # BOHLD651-08
Supplier:  Bohlender

Specifications

  • Tube ext.Ø:
    2 × max. 6 mm
  • Neck:
    2 × GL 14
  • Pk:
    1

Specifications

About this item

Blue screw cap and distributor body both made of PTFE.

  • Very good chemical resistance
  • Temperature resistant from −20 to +200 °C
  • Suitable for vacuum applications

For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with 6 mm external diameter can be inserted through the necks.