Cavity slides for diagnosis
Supplier: MARIENFELD
Certificates
About this item
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Frequently Bought Together