ContactUsLinkComponent
Diamond pen with wooden handle
Catalog # MARI6120300
Supplier:  MARIENFELD
undefined
Diamond pen with wooden handle
Catalog # MARI6120300
Supplier:  MARIENFELD
Specifications
  • Description:
    Diamond pen, wooden handle
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
For permanent labelling of microscope slides and other glass products. Indelibly engraved markings comply particularly with the guidelines of GLP.
  • Resistant to humidity, heat, cold and solvents
  • With wooden handle