Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Water, purified, sterile

Glass bottle, 90 ml vials, 10
Catalog # AX021376
Supplier:   VWR Chemicals
Water, purified, sterile
Catalog # AX021376
Supplier:  VWR Chemicals
CAS Number:  7732-18-5
Water, purified, sterile
Water, purified, sterile

Glass bottle, 90 ml vials, 10

CAS Number:  7732-18-5
Price
123,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specification Test Results

  • Aspect:
    Limpid
  • Volume (ml):
    90 ± 1,8 ml
  • Membrane filtration according to PE 2.6.1:
    Sterile after 14 days (Incubation 20-25°C & 30-35°C)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10
  • Pk Info:
    90 ml vials

Specifications