Potassium nitrate ≥97%, TECHNICAL
Plastic bucket, 5 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Nitrate of Potash
- CAS Number:7757-79-1
- Formula:KNO₃
- MDL Number:MFCD00011409
- Molecular Weight:101.1 g/mol
- Merck Index:13,07733
- Melting Pt:334 °C
- Density:2.109 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-818-8
- UN:1486
- ADR:5.1,III
- REACH:01-2119488224-35
Documents
Certificates
Search by Lot Number5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
P20_VWRC26857.368_SPEC_EN.pdf spec_VWRC26857.368
Product specification
Specifications
Specification Test Results
- Assay:Min. 97 %
Specifications
- Pack type:Plastic bucket
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



