Order Entry
Austria
Orders LinkContactUsLinkComponent
Potassium nitrate 99+% ACS

Potassium nitrate 99+% ACS

Potassium nitrate 99+% ACS
Certificates
Potassium nitrate 99+% ACS

3 Options Available Below

Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2,109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • pH:
    4.5 to 8.5 (5% soln. at 25°C)
  • Titration after Ion exchange:
    ≥99.0 %
  • Chloride (Cl):
    ≤0.002 %
  • Iodate (IO3):
    ≤5 ppm
  • Nitrite (NO2):
    ≤0.001 %
  • Phosphate (PO4):
    ≤5 ppm
  • Sulfate (SO4):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • Iron (Fe):
    ≤3 ppm
  • Magnesium (Mg):
    ≤0.002 %
  • Sodium (Na):
    ≤0.005 %
  • Heavy metals (ICP-OES):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 %
Product Family Options

Product Information