Order Entry
Austria
ContactUsLinkComponent
Sodium nitrite for synthesis, Sigma-Aldrich®
Oxidizing
Toxic
Environment
Sodium nitrite for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Oxidizing
Toxic
Environment
Sodium nitrite for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    820 g/l (20 °C)
  • Melting Point:
    280 °C (decomposition)
  • Molar Mass:
    69.00 g/mol
  • Bulk Density:
    1200 kg/m3
  • Boiling Point:
    320 °C (decomposition)
  • Density:
    2.1 g/cm3 (20 °C)
  • pH:
    9 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    510 °C
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together