Order Entry
Austria
ContactUsLinkComponent
Sodium nitrite ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Toxic
Environment
Sodium nitrite ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Oxidizing
Toxic
Environment
Sodium nitrite ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 97.0 %
  • Calcium (Ca):
    max. 0.01 %
  • Chloride (Cl):
    max. 0.005 %
  • Potassium (K):
    max. 0.005 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.01 %
  • Sulfate (SO4):
    max. 0.01 %
  • Arsenic (As):
    max. 3
  • Iron (Fe):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together