Order Entry
Austria
ContactUsLinkComponent
Sodium azide, BAKER, Laboratory Reagent, J.T.Baker®
Sodium azide, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 9099.0100
Supplier:  Avantor
CAS Number:  
Sodium azide, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 9099.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    min. 99 %
  • Alkalinity:
    max. 0.1 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.05 %
  • Loss on Drying:
    max. 0.1 %

Specification Test Results

About this item

Synonyms: Sodium triazide
 :
 :65.01 g/mol
EINECS:247-852-1
UN:1687
 :GHS06
 :GHS09
Storage Temperature:Ambient
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536
Melting Pt:275
Density:1.85 g/cm³ (20 °C)
ADR:6.1 II
Merck Index:13,08653