




Potassium permanganate, crystalline, EMPLURA®, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Merck Index:12,07824
Melting Pt:> 240 °C (dec.)
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-760-3
ADR:5.1,II
:M-Clarity™ quality level = MQ200
UN:1490
:GHS03
:GHS05
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay (manganometric):99,0 - 100,5%
- Identity:passes test
- Appearance of solution:passes test
- Water insoluble matter:≤ 0,2 %
- Chloride (Cl):≤ 0,01 %
- Sulphate (SO₄):≤ 0,03 %
Specification Test Results
Related Information
Frequently Bought Together