


Potassium permanganate 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Merck Index:12,07824
Melting Pt: Dec.
Density:2.700 g/cm³ (20 °C)
EINECS:231-760-3
UN:1490
ADR:5.1,II
:GHS07
:GHS05
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Assay:99.0 - 100.5 %
- Assay (calculated to the dried substance):99.0 - 100.5 %
- Water insoluble matter:Max. 0.2 %
- Loss on drying (on silica gel, 18h):Max. 0.2 %
- Chloride (Cl):Max. 100 ppm
- Sulfate (SO₄):Max. 500 ppm
- Appearance of the solution:complying
- Residual solvents:complying
Specification Test Results
Frequently Bought Together