


Potassium permanganate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Merck Index:12,07824
Melting Pt:> 240 °C
Density:2.7 g/cm³ (20 °C)
EINECS:231-760-3
UN:1490
ADR:5.1,II
:GHS03
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:min. 99.0 %
- Chloride and Chlorate (as Cl):max. 0.005 %
- Insoluble Matter:max. 0.2 %
- Sulfate (SO4):max. 0.02 %
Specification Test Results
Frequently Bought Together