Order Entry
Austria
ContactUsLinkComponent
Potassium iodate, Certipur® volumetric standard, secondary reference material for iodometry traceable to NIST, Supelco®
Oxidizing
Corrosive
Irritant
Potassium iodate, Certipur® volumetric standard, secondary reference material for iodometry traceable to NIST, Supelco®
Supplier:  Merck
Certificates
Oxidizing
Corrosive
Irritant
Potassium iodate, Certipur® volumetric standard, secondary reference material for iodometry traceable to NIST, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    92 g/l (25 °C)
  • Melting Point:
    560 °C decomposes
  • Molar Mass:
    214 g/mol
  • Bulk Density:
    2000 kg/m3
  • Density:
    3.98 g/cm3 (20 °C)
  • pH:
    6 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

Frequently Bought Together