Order Entry
Austria
ContactUsLinkComponent
 
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
 :  WTW
 
undefined
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
 :  WTW
 

 

  • Catalog No:
  • 662-1359
  • 662-1363
  • 662-1364
  • 662-1358
  • 662-1366
  • 662-1368
  • 662-1362
  • 662-1349
  • 662-1357
  • 662-1369
  • Model:
  • pH 7110
  • pH 7310
  • pH 7310
  • pH 7110
  • pH 7310
  • pH 7310
  • pH 7310
  • pH 7110
  • pH 7110
  • pH 7310
  • pH range:
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • −2,0...+20,0 / −2,00...+20,00 / −2,000...+19,999
  • pH accuracy:
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • pH calibration:
  • 1-, 2-, 3-point WTW technical buffer, DIN/NIST buffer
  • 1-, 2-, 3-, 4-, 5-point, WTW technical buffer, DIN/NIST 20 additional buffer sets
  • 1-, 2-, 3-, 4-, 5-point, WTW technical buffer, DIN/NIST 20 additional buffer sets
  • 1-, 2-, 3-point WTW technical buffer, DIN/NIST buffer
  • 1-, 2-, 3-, 4-, 5-point, WTW technical buffer, DIN/NIST 20 additional buffer sets
  • 1-, 2-, 3-, 4-, 5-point, WTW technical buffer, DIN/NIST 20 additional buffer sets
  • 1-, 2-, 3-, 4-, 5-point, WTW technical buffer, DIN/NIST 20 additional buffer sets
  • 1-, 2-, 3-point WTW technical buffer, DIN/NIST buffer
  • 1-, 2-, 3-point WTW technical buffer, DIN/NIST buffer
  • 1-, 2-, 3-, 4-, 5-point, WTW technical buffer, DIN/NIST 20 additional buffer sets
  • mV range:
  • ±1200,0 / ±2000
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • ±1200,0 / ±2000
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • ±1200,0 / ±2000
  • ±1200,0 / ±2000
  • ±1200,0 / ±2500
  • mV accuracy:
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • Temperature range:
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Display:
  • 7-segment LCD
  • Graphic display with backlight
  • Graphic display with backlight
  • 7-segment LCD
  • Graphic display with backlight
  • Graphic display with backlight
  • Graphic display with backlight
  • 7-segment LCD
  • 7-segment LCD
  • Graphic display with backlight
  • Power supply:
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Universal power supply, 4× 1,5 V AA or 4× 1,2 V NiMH-rechargeable battery
  • Data logging capacity:
  • -
  • 500 manually / 5000 automatic
  • 500 manually / 5000 automatic
  • -
  • 500 manually / 5000 automatic
  • 500 manually / 5000 automatic
  • 500 manually / 5000 automatic
  • -
  • -
  • 500 manually / 5000 automatic
  • Connections:
  • -
  • Mini USB
  • Mini USB
  • -
  • Mini USB
  • Mini USB
  • Mini USB
  • -
  • -
  • Mini USB
  • Outputs:
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana
  • DIN or BNC / 4 mm banana

 

Frequently Bought Together