Order Entry
Austria
ContactUsLinkComponent
Sodium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31535.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31535.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Merck Index:14,08599