Order Entry
Austria
ContactUsLinkComponent
 
Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
 
Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
 

 

  • Assay (argentometric; calculated on dried substance):
    99.5 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Acidity or alkalinity:
    passes test
  • Bromide (Br):
    ≤ 0.005 %
  • Ferrocyanides:
    passes test
  • Iodide (I):
    passes test
  • Nitrite (NO₂) (UV-absorption 354nm; 1cm; 10%; water):
    ≤ 0.01
  • Phosphate (PO₄):
    ≤ 0.0025 %
  • Sulfate (SO₄):
    ≤ 0.0200 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • As (Arsenic):
    ≤ 0.0001 %
  • Ba (Barium):
    passes test
  • Ca (Calcium):
    ≤ 0.002 %
  • K (Potassium):
    ≤ 0.01 %
  • Fe (Iron):
    ≤ 0.0002 %
  • Magnesium and alkaline-earth metals (as Ca):
    ≤ 0.0100 %
  • Residual solvents (ICH Q3C):
    excluded by the manufacturing process
  • Loss on drying (105 °C, 2 h):
    ≤ 0.5 %

 

Frequently Bought Together