Order Entry
Austria
ContactUsLinkComponent
Sodium chloride, Certipur® volumetric standard, secondary reference material, Supelco®
Sodium chloride, Certipur® volumetric standard, secondary reference material, Supelco®
Catalog # 1.02406.0080
Supplier:  Merck
CAS Number:  
Sodium chloride, Certipur® volumetric standard, secondary reference material, Supelco®
Catalog # 1.02406.0080
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    80 g
  • Seq:
    3392

Specifications

Specification Test Results

  • Solubility:
    358 g/l (20 °C)
  • Melting Point:
    801 °C
  • Molar Mass:
    58.44 g/mol
  • Bulk Density:
    1140 kg/m3
  • Boiling Point:
    1461 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (865 °C)
  • Density:
    2.17 g/cm3 (20 °C)
  • pH:
    6 - 9 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :58.44 g/mol
Merck Index:14,08599
Boiling Pt:1461 °C (1013 hPa)
Melting Pt:801 °C
Density:2.17 g/cm3 (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477