Order Entry
Austria
ContactUsLinkComponent
 
Sodium chloride, crystals, BAKER ANALYZED®, J.T.Baker®
 :  Avantor
 
Sodium chloride, crystals, BAKER ANALYZED®, J.T.Baker®
 :  Avantor
 

 

  • Assay:
    min. 99.5 %
  • Barium (Ba):
    max. 0.002 %
  • Bromide (Br):
    max. 0.01 %
  • Calcium (Ca):
    max. 0.002 %
  • Iodide (I):
    max. 0.005 %
  • Potassium (K):
    max. 0.01 %
  • Magnesium (Mg):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    5.0 - 8.0
  • Sulfate (SO4):
    max. 0.01 %
  • Arsenic (As):
    max. 3
  • Iron (Fe):
    max. 3
  • Heavy Metals (as Pb):
    max. 5
  • Phosphate (PO4):
    max. 5

 

Frequently Bought Together