Lithium chloride
MOLE22094921-500G
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
:
:42.39 g/mol
:13,05550
:1382 °C (1013 hPa)
:614 °C
:2.07 g/cm³ (20 °C)
:Ambient
:231-212-3
:GHS07
:7447-41-8
:LiCl
:MFCD00011078