Order Entry
Austria
ContactUsLinkComponent
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    832 g/l (20 °C)
  • Melting Point:
    614 °C
  • Molar Mass:
    42.39 g/mol
  • Bulk Density:
    530 kg/m3
  • Boiling Point:
    1360 °C (1013 hPa)
  • Vapor Pressure:
    1.33 hPa (547 °C)
  • Density:
    2.07 g/cm3 (20 °C)
  • pH:
    6 (50 g/l, H2O, 20 °C)

 

Frequently Bought Together