Order Entry
Austria
ContactUsLinkComponent
Potassium chloride ≥99%, GPR RECTAPUR®
Potassium chloride ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Potassium chloride ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together