Order Entry
Austria
ContactUsLinkComponent
 
Potassium chloride ≥97%, TECHNICAL
Catalog # 26752.366
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium chloride ≥97%, TECHNICAL
Catalog # 26752.366
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

About this item

 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360