Order Entry
Austria
ContactUsLinkComponent
 
Potassium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
Potassium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    347 g/l (20 °C)
  • Melting Point:
    773 °C
  • Molar Mass:
    74.55 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    1413 °C (1013 hPa)
  • Density:
    1.98 g/cm3 (20 °C)
  • pH:
    5.5 - 8.0 (50 g/l, H2O, 25 °C)

 

Frequently Bought Together