Order Entry
Austria
ContactUsLinkComponent
 
Potassium chloride 99.5-100.5%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
 
Potassium chloride 99.5-100.5%, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
 

 

  • Assay:
    Min. 99.5 %
  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Loss on drying (105°C, 3h):
    Max. 0.2 %
  • pH (5 %, 25 °C):
    5-8
  • Acid. or alk. react. impurities:
    complying
  • Free acid (as HCl):
    Max. 0.003 %
  • Free alkali (as KOH):
    Max. 0.003 %
  • Aluminium (Al):
    Max. 1 ppm
  • Barium (Ba):
    Max. 0.001 %
  • Calcium (Ca):
    Max. 0.001 %
  • Iron (Fe):
    Max. 2 ppm
  • Magnesium (Mg):
    Max. 0.0005 %
  • Sodium (Na):
    Max. 0.02 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Mg, earth alkali metals (as Ca):
    Max. 200 ppm
  • Bromide (Br):
    Max. 0.01 %
  • Iodide (I):
    Max. 0.002 %
  • Iodide (I) test:
    complying
  • Phosphate (PO₄):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 20 ppm
  • Total N:
    Max. 0.001 %
  • Appearance of the solution:
    complying

 

 

Frequently Bought Together