Cesium chloride ≥99.99%, Gen-Apex, high purity for molecular biology
Catalog # 33605.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 99.99 %
- Colouration (0,25 mol/l; water):Max. 10 APHA
- Heavy metals (as Pb):Max. 20 ppm
- Transmittance (260 nm) (50 %):Min. 99 %
- Transmittance (280 nm) (50 %):Min. 99 %
Specification Test Results
About this item
:
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:646 °C
Density:3.7808 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-600-2
CAS Number:7647-17-8
Formula:CsCl
MDL Number:MFCD00010955