Order Entry
Austria
ContactUsLinkComponent
Cesium chloride ≥99.5%, AnalaR NORMAPUR® analytical reagent
Cesium chloride ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Cesium chloride ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Total N (Nitrogen):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Al (Aluminium):
    Max. 5 ppm
  • Cu (Copper):
    Max. 3 ppm
  • Fe (Iron):
    Max. 3 ppm
  • K (Potassium):
    Max. 20 ppm
  • Li (Lithium):
    Max. 2 ppm
  • Mg (Magnesium):
    Max. 5 ppm
  • Na (Sodium):
    Max. 20 ppm
  • Pb (Lead):
    Max. 1 ppm
  • Rb (Rubidium):
    Max. 200 ppm
  • Zn (Zinc):
    Max. 3 ppm

Specification Test Results

Frequently Bought Together