Order Entry
Austria
ContactUsLinkComponent
Cesium chloride analytical reagent, Supelco®
Cesium chloride analytical reagent, Supelco®
Supplier:  Merck
Certificates
Cesium chloride analytical reagent, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    1860 g/l (20 °C)
  • Melting Point:
    646 °C
  • Molar Mass:
    168.36 g/mol
  • Bulk Density:
    1800 kg/m3
  • Boiling Point:
    1382 °C
  • Density:
    3.97 g/cm3 (20 °C)
  • pH:
    7 - 9 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together