Order Entry
Austria
ContactUsLinkComponent
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Certificates
undefined
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Certificates

Specification Test Results

  • Appearance:
    Colorless to white Crystalline powder or crystals
  • UV:
    (50 % in water) 1 cm cell
  • Purity:
    ≥99.94 % (metals basis)
  • Insoluble matter:
    ≤25 ppm (in water)

Specification Test Results

Related Information

Frequently Bought Together