Order Entry
Austria
ContactUsLinkComponent
Sodium hydrogen carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Sodium hydrogen carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Sodium hydrogen carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    96 g/l (20 °C)
  • Melting Point:
    270 °C (decomposition)
  • Molar Mass:
    84.01 g/mol
  • Bulk Density:
    1000 kg/m3
  • Vapor Pressure:
    8.3 hPa (30 °C)
  • Density:
    2.22 g/cm3 (20 °C)
  • pH:
    8.6 (50 g/l, H2O, 20 °C)

Specification Test Results

Frequently Bought Together