Order Entry
Austria
ContactUsLinkComponent
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Certificates
undefined
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Food GMP Manufactured Product:
  • GMP Manufactured Product:
  • Meets E.P. Chemical Specifications:
  • Meets U.S.P Requirements:
  • Meets J.P. Chemical Specifications:
  • Meets B.P. Chemical Specifications:
  • Meets F.C.C. Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • Bulk Food Chemical:
  • USP - Assay (NaHCO₃) (dried basis):
    99.0 - 100.5 %
  • USP - Limit of Ammonia:
    ≤ 20 ppm
  • USP - Limit of Sulfur Compounds:
    ≤ 0.015 %
  • USP - Arsenic (As):
    ≤ 2 ppm
  • USP - Normal Carbonate:
    Passes Test
  • USP - Chloride (Cl):
    ≤ 0.015 %
  • USP - Identification A (Sodium):
    Passes Test
  • USP - Identification B (Bicarbonate):
    Passes Test
  • USP - Insoluble Substances:
    Passes Test
  • USP - Loss on Drying:
    ≤ 0.25 %
  • EP/BP - Assay (NaHCO₃):
    99.0 - 101.0 %
  • EP/BP - Ammonium (NH₄):
    ≤ 20 ppm
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Calcium (Ca):
    ≤ 100 ppm
  • EP/BP - Carbonate (pH):
    ≤ 8.6
  • EP/BP - Chloride (Cl):
    ≤ 150 ppm
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 20 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 150 ppm
  • JP - Assay (NaHCO₃):
    99.0 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - pH (1 in 20):
    7.9 - 8.4
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.040 %
  • JP - Carbonate (CO₃):
    Passes Test
  • JP - Ammonium (NH₄):
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 5 ppm
  • JP - Arsenic (As):
    ≤ 2 ppm
  • FCC - Assay (NaHCO₃) (dried basis):
    99.0 - 100.5 %
  • FCC - Insoluble Substances:
    Passes Test
  • FCC - Lead (Pb):
    ≤ 2 mg/kg
  • FCC - Limit of Ammonia (NH₃):
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤ 5.0
  • No Class 1,2,3 or other solvents are used or produced in the:
    ≤ 5.0

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together