
Sodium carbonate, Certipur® volumetric standard, secondary reference material, Supelco®
Catalog # 1.02405.0080
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:80 g
- Seq:3392
Specifications
Specification Test Results
- Solubility:220 g/l (20 °C)
- Melting Point:854 °C
- Molar Mass:105.99 g/mol
- Bulk Density:1100 kg/m3
- Boiling Point:1600 °C (decomposition)
- Density:2.53 g/cm3 (20 °C)
- pH:11.5 (50 g/l, H2O, 25 °C)
Specification Test Results
About this item
:
:105.99 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494