Order Entry
Austria
ContactUsLinkComponent
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Supplier:  Thermo Orion
Certificates
undefined
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
Supplier:  Thermo Orion
Limited Time Sale
Select items below are currently on sale

Specifications

  • Catalog No:
  • 662-1443
  • ORIOSTARA2116
  • 662-1442
  • ORIOSTARA2114
  • ORIOSTARA2117
  • ORIOSTARA2119
  • Model:
  • A211
  • A211
  • A211
  • A211
  • A211
  • A211
  • pH range:
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • Temperature range:
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery type:
  • 4×AA (sold separately)
  • 4×AA (sold separately)
  • 4×AA (sold separately)
  • 4×AA (sold separately)
  • 4×AA (sold separately)
  • 4×AA (sold separately)
  • Battery life:
  • 800 hours
  • 800 hours
  • 800 hours
  • 800 hours
  • 800 hours
  • 800 hours
  • Power supply:
  • AC adapter or 4 AA batteries (800 hours battery life)
  • AC adapter or 4 AA batteries (800 hours battery life)
  • AC adapter or 4 AA batteries (800 hours battery life)
  • AC adapter or 4 AA batteries (800 hours battery life)
  • AC adapter or 4 AA batteries (800 hours battery life)
  • AC adapter or 4 AA batteries (800 hours battery life)
  • Compliance:
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • Operating temperature range:
  • 5...45 °C
  • 5...45 °C
  • 5...45 °C
  • 5...45 °C
  • 5...45 °C
  • 5...45 °C
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 2000
  • 2000
  • 2000
  • 2000
  • 2000
  • 2000
  • Memory:
  • Non volatile memory preserves data log, calibration log and meter settings
  • Non volatile memory preserves data log, calibration log and meter settings
  • Non volatile memory preserves data log, calibration log and meter settings
  • Non volatile memory preserves data log, calibration log and meter settings
  • Non volatile memory preserves data log, calibration log and meter settings
  • Non volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • Connections:
  • BNC, reference pin, 8-pin mini-DIN
  • BNC, reference pin, 8-pin mini-DIN
  • BNC, reference pin, 8-pin mini-DIN
  • BNC, reference pin, 8-pin mini-DIN
  • BNC, reference pin, 8-pin mini-DIN
  • BNC, reference pin, 8-pin mini-DIN
  • Outputs:
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • Input:
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Calibration:
  • pH with calibration editing option, relative mV, ORP and temperature
  • pH with calibration editing option, relative mV, ORP and temperature
  • pH with calibration editing option, relative mV, ORP and temperature
  • pH with calibration editing option, relative mV, ORP and temperature
  • pH with calibration editing option, relative mV, ORP and temperature
  • pH with calibration editing option, relative mV, ORP and temperature
  • Calibration points:
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • W×D×H:
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • Weight:
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg
  • 0,9 kg

Specifications

Electrodes

Product Information

Frequently Bought Together