Orders LinkContactUsLinkComponent
VWR®, Refrigerated Circulators | Avantor
VWR®, Refrigerated Circulators
Supplier:   VWR Collection
VWR®, Refrigerated Circulators
Supplier:  VWR Collection
Certificates
VWR®, Refrigerated Circulators
VWR®, Refrigerated Circulators
15 Options Available Below
Product Details & Documents
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both are easy to navigate with large, intuitive displays and multiple communication options, including USB-A and B, RS232/485, Ethernet and external temperature probe. The reservoir drain can be accessed by removing the front panel. All models feature user-adjustable, high temperature safety cut-off points, as well as over-temperature protection. The surface is cool to the touch, even when operating at high temperatures.
  • Working temperatures from −40 to +200 °C with stability of ±0,01 °C (except 45 litre models with maximum 135 °C)
  • Variable speed pressure/suction pump with external circulation and temperature control capability
  • 180º rotating controllers, lid stowing system, chemically resistant deck and environmental control system reduces noise, increases refrigeration efficiency, and lowers energy consumption
  • Cooling technology regulates the amount of cooling required, saving energy while providing rapid cooling and precise control at elevated temperatures
  • Event scheduling (time and date), real time clock and temperature trends for up to 10 days with Programmable models
  • Selectable home screens and on-screen help
  • Automatic and/or user-adjustable performance optimisation
Advanced Digital models feature an intuitive 9,5 cm display with touch pad control, single-point calibration, menus and prompts in four languages: French, German, Spanish and English.
Advanced Programmable models have an intuitive 10,9 cm display, five-point calibration capability, menus and prompts in six languages: French, German, Spanish, English, Chinese and Arabic, plus time/temperature programming (10×100-step programs).
Zertifizierung: Complies with DIN 12876-1, Safety Class III
Maximum pump capacities: 16,7 L/min, 250 mbar, 12,2 L/min suction
Lieferumfang: Supplied with a reservoir cover, bypass tubing, male inlet and outlet adapters for 47, 63 and 95 mm tubing, ¼" to M16 adapters are also included. For probes and accessories, please visit website or contact your local Avantor sales office.
Achtung! Bitte beachten: Cooling capacity at 20 °C
Product Family Options
Models with Advanced Digital temperature controller
Product Information
  • Type
    Temperature Range
    Temp. stability
    Heating capacity
    Cooling capacity
    Capacity
    W×D×H
    Availability
    Price
  • AD7LR-20
    −20…+200 °C
    ±0,01 °C
    2,2 kW
    0,2 kW
    7 l
    411×589×411 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 950,00 €
    6 950,00 €
    1 * 1 ST
       6 950,00 €
  • AD07R-20
    −20…+200 °C
    ±0,01 °C
    2,2 kW
    0,2 kW
    7 l
    221×541×617 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 060,00 €
    6 060,00 €
    1 * 1 ST
       6 060,00 €
  • AD07R-40
    −40…+200 °C
    ±0,01 °C
    2,2 kW
    0,36 kW
    7 l
    221×541×617 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 100,00 €
    6 100,00 €
    1 * 1 ST
       6 100,00 €
  • AD15R-30
    −30…+200 °C
    ±0,01 °C
    2,2 kW
    0,915 kW
    15 l
    368×569×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 350,00 €
    7 350,00 €
    1 * 1 ST
       7 350,00 €
  • AD15R-40
    −40…+200 °C
    ±0,01 °C
    2,2 kW
    1,0 kW
    15 l
    368×569×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 620,00 €
    7 620,00 €
    1 * 1 ST
       7 620,00 €
  • AD20R-30
    −30…+200 °C
    ±0,01 °C
    2,2 kW
    0,915 kW
    20 l
    419×610×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 680,00 €
    7 680,00 €
    1 * 1 ST
       7 680,00 €
  • AD28R-30
    −30…+200 °C
    ±0,01 °C
    2,2 kW
    0,915 kW
    28 l
    457×672×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 340,00 €
    8 340,00 €
    1 * 1 ST
       8 340,00 €
  • AD45R-20
    −25…+135 °C
    ±0,01 °C
    2,2 kW
    1,4 kW
    45 l
    568×909×1070 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14 730,00 €
    14 730,00 €
    1 * 1 ST
       14 730,00 €
Models with Advanced Programmable temperature controller
Product Information
  • Type
    Temperature Range
    Temp. stability
    Heating capacity
    Cooling capacity
    Capacity
    W×D×H
    Availability
    Price
  • AP07R-20
    −20...+200 °C
    ±0,01 °C
    2,2 kW
    0,2 kW
    7 L
    221×541×617 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 430,00 €
    6 430,00 €
    1 * 1 ST
       6 430,00 €
  • AP07R-40
    −40…+200 °C
    ±0,01 °C
    2,2 kW
    0,36 kW
    7 L
    221×541×617 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 000,00 €
    8 000,00 €
    1 * 1 ST
       8 000,00 €
  • AP15R-30
    −30…+200 °C
    ±0,01 °C
    2,2 kW
    0,915 kW
    15 L
    368×569×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 960,00 €
    7 960,00 €
    1 * 1 ST
       7 960,00 €
  • AP15R-40
    −40…+200 °C
    ±0,01 °C
    2,2 kW
    1,0 kW
    15 L
    368×569×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 500,00 €
    8 500,00 €
    1 * 1 ST
       8 500,00 €
  • AP20R-30
    −30…+200 °C
    ±0,01 °C
    2,2 kW
    0,915 kW
    20 L
    419×610×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 870,00 €
    7 870,00 €
    1 * 1 ST
       7 870,00 €
  • AP28R-30
    −30…+200 °C
    ±0,01 °C
    2,2 kW
    0,915 kW
    28 L
    457×672×683 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 600,00 €
    8 600,00 €
    1 * 1 ST
       8 600,00 €
  • AP45R-20
    −25…+135 °C
    ±0,01 °C
    2,2 kW
    1,4 kW
    45 L
    568×909×1070 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15 910,00 €
    15 910,00 €
    1 * 1 ST
       15 910,00 €
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Digital to analogue adapter
    Digital to analogue adapter
    461-0417
    VWR Bath and refrigerated circulators, P and D models
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • PT100 probe with 0,6 m cable
    PT100 probe with 0,6 m cable
    462-0271
    Log in for availability
    1 * 1 ST - 487,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PT100 probe with 7,6 m cable
    PT100 probe with 7,6 m cable
    462-0273
    Log in for availability
    1 * 1 ST - 730,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PT100 probe with 15 m cable
    PT100 probe with 15 m cable
    462-0274
    Log in for availability
    1 * 1 ST - 1 080,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PT100 probe with 3 m cable
    PT100 probe with 3 m cable
    462-0272
    Log in for availability
    1 * 1 ST - 540,00 €