
Potassium bromide, EMPROVE® EVOLVE Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7758-02-3
:
Formula:KBr
MDL Number:MFCD00011358
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
:119 g/mol
EINECS:231-830-3
:GHS07
Storage Temperature:Ambient
Related Information
MERP1.04900.1000_TDS_EN ProductPage_104900 ProductPage_104900
Product specification
msds_104900
SDS
Frequently Bought Together