


di-Lithium tetraborate analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:12007-60-2
:
Formula:Li₂B₄O₇
MDL Number:MFCD00011083
:169.12 g/mol
Merck Index:14,05524
Melting Pt:918 °C
Density:1.4 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:234-514-3
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:50 g/l (20 °C)
- Melting Point:930 °C
- Molar Mass:169.12 g/mol
- Bulk Density:330 kg/m3
- pH:9.1 (100 g/l, H2O, 20 °C) (slurry)
Specification Test Results
Frequently Bought Together