
di-Lithium tetraborate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12007-60-2
:
Formula:Li₂B₄O₇
MDL Number:MFCD00011083
:169.12 g/mol
Merck Index:14,5524
Melting Pt:930 °C
Density:2.4
EINECS:234-514-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together