Order Entry
Austria
ContactUsLinkComponent
Sodium tetrachloroaurate dihydrate, TECHNICAL
Irritant
Sodium tetrachloroaurate dihydrate, TECHNICAL
Catalog # 26069.081
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Sodium tetrachloroaurate dihydrate, TECHNICAL
Catalog # 26069.081
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :397.8 g/mol
EINECS:239-241-3
UN:3260
ADR:8,III
 :GHS07
CAS Number:13874-02-7
Formula:NaAuCl₄·2H₂O
MDL Number:MFCD00149162