

Lithium aluminum hydride, Sigma-Aldrich®
: Merck
:16853-85-3
:
:H₄AlLi
:MFCD00011075
:37.95 g/mol
:13,00348
:125 °C
:0.91 g/cm³ (20 °C)
:Ambient
:240-877-9
:1410
:4.3,I
:GHS02
:GHS05
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals