Order Entry
Austria
ContactUsLinkComponent
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Flammable
Health
Irritant
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue:
    Max. 3 ppm
  • Water:
    Max. 30 ppm

Specification Test Results

Related Information

Frequently Bought Together