Order Entry
Austria
ContactUsLinkComponent
1,4-Dioxane, dried ≥99.5% (by GC) stabilised, Puriss. p.a. (≤0.005% H₂O), Burdick & Jackson™
Flammable
Health
Irritant
1,4-Dioxane, dried ≥99.5% (by GC) stabilised, Puriss. p.a. (≤0.005% H₂O), Burdick & Jackson™
Catalog # 34944-1L
CAS Number:  
undefined
Flammable
Health
Irritant
1,4-Dioxane, dried ≥99.5% (by GC) stabilised, Puriss. p.a. (≤0.005% H₂O), Burdick & Jackson™
Catalog # 34944-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Congealing - Freezing Point:
    Min. 11 °C
  • Boiling range:
    100 - 102 °C
  • Density (D 20/4):
    1.031 - 1.034
  • Refractive index (n 20/D):
    1.4210 - 1.4240
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.005 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.001 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • Acetaldehyde:
    Max. 0.005 %
  • Carbonyl Compounds (as HCHO):
    Max. 0.01 %
  • Peroxides (as H₂O₂):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03330
Boiling Pt:100-102 °C (1.013 hPa)
Boiling Pt:100…102 °C (1.013 hPa)
Melting Pt:12 °C
Density:1.031…1.034 g/cm³ (20 °C)
Flash Pt:22.5 %(V)
Storage Temperature:Ambient
EINECS:204-661-8
UN:1165
ADR:3,II
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ~25 mg/L
 :GHS02
 :GHS08
 :GHS07
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571